missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexylchlorophosphine, 98+%

Product Code. 11327768
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327768 1 g 1g
11317768 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327768 Supplier Thermo Scientific Alfa Aesar Supplier No. 033571.03
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16523-54-9 | C12H22ClP | 232.73 g/mol

Dicyclohexylchlorophosphine is used as pharmaceutical intermediate. It is also used in the synthesis of various phosphines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16523-54-9
Molecular Formula C12H22ClP
Molecular Weight (g/mol) 232.73
MDL Number MFCD00014294
InChI Key AKJFBIZAEPTXIL-UHFFFAOYSA-N
Synonym chlorodicyclohexylphosphine, dicyclohexylchlorophosphine, dicyclohexylphosphinous chloride, dicyclohexylphosphine chloride, chlorodicyclohexyl phosphine, phosphinous chloride, dicyclohexyl, clpcy2, pcy2cl, chlorodicyclohexvlphosphine, dicyclohexylchlorophosphane
PubChem CID 2734135
SMILES ClP(C1CCCCC1)C1CCCCC1

Specifications

Color Colorless to Yellow
Density 1.054 g/mL
Boiling Point 97°C (0.1 mmHg)
Flash Point >110°C (230°F)
Odor Acrid
Linear Formula (C6H11)2PCl
Refractive Index 1.533
Quantity 1 g
UN Number UN3265
Sensitivity Air and moisture sensitive
Solubility Information Reacts violently with water.
IUPAC Name chloro(dicyclohexyl)phosphane
Formula Weight 232.73
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Dicyclohexylchlorophosphine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.