missing translation for 'onlineSavingsMsg'
Learn More

Dicyclopentadiene, 90+%, stab. with 4-tert-butylcatechol

Product Code. 11460224
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460224 500 mL 500mL
11470224 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460224 Supplier Thermo Scientific Alfa Aesar Supplier No. A13643.AP
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 77-73-6 | C10H12 | 132.206 g/mol

Dicyclopentadiene is used in resins, particularly unsaturated polyester resins. It plays a major role in inks, adhesives and paints. It is also used as a monomer in polymerization reactions. It is a precursor for the preparation of endo-tetrahydrodicyclopentadiene, which reacts with aluminum chloride at higher temperature to give adamantine. It undergoes a retro-Diels-Alder reaction to yield cyclopentadiene, which acts as ligand in inorganic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicyclopentadiene is used in resins, particularly unsaturated polyester resins. It plays a major role in inks, adhesives and paints. It is also used as a monomer in polymerization reactions. It is a precursor for the preparation of endo-tetrahydrodicyclopentadiene, which reacts with aluminum chloride at higher temperature to give adamantine. It undergoes a retro-Diels-Alder reaction to yield cyclopentadiene, which acts as ligand in inorganic chemistry.

Solubility
Immiscible with water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-73-6
Molecular Formula C10H12
Molecular Weight (g/mol) 132.206
MDL Number MFCD00078246
InChI Key HECLRDQVFMWTQS-UHFFFAOYSA-N
Synonym dicyclopentadiene, biscyclopentadiene, cyclopentadiene dimer, bicyclopentadiene, dicyklopentadien, dimer cyklopentadienu, dcpd, 4,7-methano-1h-indene, 3a,4,7,7a-tetrahydro, 3a,4,7,7a-tetrahydro-1h-4,7-methanoindene, 1,3-cyclopentadiene, dimer
PubChem CID 6492
ChEBI CHEBI:34695
SMILES C1C=CC2C1C3CC2C=C3

Specifications

Density 0.982
Boiling Point 171°C to 173°C
Flash Point 26°C (78°F)
Refractive Index 1.51
Quantity 500 mL
UN Number UN2048
Beilstein 1904092
Merck Index 14,2739
Solubility Information Immiscible with water.
Formula Weight 132.21
Percent Purity ≥90%
Chemical Name or Material Dicyclopentadiene, stabilized with 4-tert-butylcatechol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dicyclopentadiene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Dicyclopentadiene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.