missing translation for 'onlineSavingsMsg'
Learn More

Dicyclopentadiene, 95%, stabilized with 100-200 ppm 4-tert-Butylcatechol

Product Code. 10724841 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
100mL
2.5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10724841 2.5 L Glass bottle 2.5 L
10636162 100 mL Glass bottle 100mL
10377370 1 L Glass bottle 1 L
11954431 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10724841 Supplier Thermo Scientific Acros Supplier No. 150760025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 77-73-6 | C10H12 | 132.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-73-6 , 98-29-3
Molecular Formula C10H12
Molecular Weight (g/mol) 132.2
InChI Key HECLRDQVFMWTQS-UHFFFAOYSA-N
Synonym dicyclopentadiene, biscyclopentadiene, cyclopentadiene dimer, bicyclopentadiene, dicyklopentadien, dimer cyklopentadienu, dcpd, 4,7-methano-1h-indene, 3a,4,7,7a-tetrahydro, 3a,4,7,7a-tetrahydro-1h-4,7-methanoindene, 1,3-cyclopentadiene, dimer
PubChem CID 6492
ChEBI CHEBI:34695
SMILES C1C=CC2C1C3CC2C=C3

Specifications

Melting Point 11.0°C to 13.0°C
Density 0.9820g/mL
Boiling Point 170.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 91% min. (endo-Dicyclopentadiene) (GC)
Packaging Glass bottle
Refractive Index 1.5100 to 1.5120
Quantity 2.5 L
Beilstein 05,II,391
Merck Index 14,2739
Specific Gravity 0.982
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in ethanol,diethyl ether
Stabilizer 100 to 200ppm p-tert.-butylcatechol
Formula Weight 132.2
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Dicyclopentadiene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.