missing translation for 'onlineSavingsMsg'
Learn More

Dicyclopropylmethanol, 97%

Product Code. 11391407
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11391407 1 g 1g
11301417 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391407 Supplier Thermo Scientific Alfa Aesar Supplier No. L01816.03
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 14300-33-5 | C7H12O | 112.17 g/mol

Dicyclopropylmethanol is used as protecting group reagent for carboxylic acids; the formed esters are very mildly and selectively cleaved.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicyclopropylmethanol is used as protecting group reagent for carboxylic acids; the formed esters are very mildly and selectively cleaved.

Solubility
Sparingly soluble in water.

Notes
Store at 2-8°C. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14300-33-5
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00019249
InChI Key PIXLZMHERIHLJL-UHFFFAOYSA-N
Synonym dicyclopropyl carbinol, dicyclopropylmethyl alcohol, dicyclopropylmethan-1-ol, cyclopropanemethanol, a-cyclopropyl, cyclopropanemethanol, .alpha.-cyclopropyl, dicyclopropyl-methanol, acmc-20ala1, alpha-cyclopropylcyclopropanemethanol
PubChem CID 84336
IUPAC Name dicyclopropylmethanol
SMILES OC(C1CC1)C1CC1

Specifications

Boiling Point 99°C to 100°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.465
Quantity 1 g
Beilstein 2203215
Solubility Information Sparingly soluble in water.
Formula Weight 112.17
Percent Purity 97%
Chemical Name or Material Dicyclopropylmethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.