missing translation for 'onlineSavingsMsg'
Learn More

Didanosine, MedChemExpress

Product Code. 16438438
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438438 10 mg 10mg
16424317 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16438438 Supplier Medchem Express Supplier No. HYB024910mg
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Didanosine(Videx) is a reverse transcriptase inhibitor with an IC50 of 0.49 μM.

Didanosine(Videx) is a reverse transcriptase inhibitor with an IC50 of 0.49 μM.

Specifications

Chemical Name or Material Didanosine
CAS 69655-05-6
Color White
Purity Grade Notes Research
Molecular Formula C10H12N4O3
Quantity 10 mg
Synonym 2',3'-Dideoxyinosine ddI
Solubility Information DMSO : 62.5 mg/mL (264.57 mM; Need ultrasonic)
SMILES OC[C@@H]1CC[C@H](N2C=NC3=C2N=CN=C3O)O1
Molecular Weight (g/mol) 236.23
Formula Weight 236.23
Percent Purity 99.75%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.