missing translation for 'onlineSavingsMsg'
Learn More

Didodecyldimethylammonium Chloride, TRC

Product Code. 17984765
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17984765 10 g 10g
17943865 1 g 1g
17953745 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17984765 Supplier Toronto Research Chemicals Supplier No. TRCD44176010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3401-74-9
Molecular Formula C26 H56 N . Cl
Molecular Weight (g/mol) 418.18
Synonym 1-Dodecanaminium, N-dodecyl-N,N-dimethyl-, chloride (9CI), Ammonium, didodecyldimethyl-, chloride (8CI), Didodecyldimethylammonium chloride (6CI,7CI), AQ 212, Aliquat 204, Dilauryldimethylammonium chloride, Dimethyldidodecylammonium chloride, Dimethyldilaurylammonium chloride, Isothan DL, Isothan DL 1, Quartamin D 24E, Quaternium 47, 1-Dodecanaminium, N-dodecyl-N,N-dimethyl-, chloride (9CI), Ammonium, didodecyldimethyl-, chloride (8CI), Didodecyldimethylammonium chloride (6CI,7CI), AQ 212, Aliquat 204, Dilauryldimethylammonium chloride, Dimethyldidodecylammonium chloride, Dimethyldilaurylammonium chloride, Isothan DL, Isothan DL 1, Quartamin D 24E, Quaternium 47
IUPAC Name didodecyl(dimethyl)azanium;chloride
SMILES [Cl-].CCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C26H56N.ClH/c1-5-7-9-11-13-15-17-19-21-23-25-27(3,4)26-24-22-20-18-16-14-12-10-8-6-2;/h5-26H2,1-4H3;1H/q+1;/p-1
Formula Weight 417.4101
Percent Purity >95%
Chemical Name or Material Didodecyldimethylammonium Chloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.