missing translation for 'onlineSavingsMsg'
Learn More

Diethanolamine, TRC

Product Code. 30132471
Change view
Click to view available options
Quantity:
5 g
25 g
500 g
Unit Size:
25g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132471 500 g 500g
30137505 25 g 25g
17945005 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132471 Supplier Toronto Research Chemicals Supplier No. TRCD441900500G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-42-2
Molecular Formula C4 H11 N O2
Molecular Weight (g/mol) 105.14
Synonym 2,2'-Iminobisethanol, Ethanol, 2,2'-iminodi- (8CI), 2,2'-Azanediyldiethanol, 2,2'-Dihydroxydiethylamine, 2,2'-Iminobis[ethanol], 2,2'-Iminodi-1-ethanol, 2,2'-Iminodiethanol, 2-[(2-Hydroxyethyl)amino]ethanol, Bis(2-hydroxyethyl)amine, Bis(hydroxyethyl)amine, DEA, DEA 80, DEAO-LF, DEOA LF, Dabco DEOA-LF, Di(2-hydroxyethyl)amine, Di(β-hydroxyethyl)amine, Diethanolamine, Diethanolamine 80, Diolamine, Iminodiethanol, N,N-Bis(2-hydroxyethyl)amine, N,N-Di(2-hydroxyethyl)amine, N,N-Diethanolamine, NSC 4959, Niax DEOA-LF, Tegoamin DEOA 85
IUPAC Name 2-(2-hydroxyethylamino)ethanol
SMILES OCCNCCO

Specifications

Quantity 500 g
InChI Formula InChI=1S/C4H11NO2/c6-3-1-5-2-4-7/h5-7H,1-4H2
Formula Weight 105.079
Chemical Name or Material Diethanolamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.