missing translation for 'onlineSavingsMsg'
Learn More

Diethoxydiphenylsilane, 98%

Product Code. 11475826
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475826 50 g 50g
11465826 10 g 10g
11485826 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475826 Supplier Thermo Scientific Alfa Aesar Supplier No. B22204.18
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2553-19-7 | C16H20O2Si | 272.42 g/mol

Reagent for preparation of diphenylsilylene derivatives of diols by exchange.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2553-19-7
Molecular Formula C16H20O2Si
Molecular Weight (g/mol) 272.42
MDL Number MFCD00015126
InChI Key ZZNQQQWFKKTOSD-UHFFFAOYSA-N
Synonym diphenyldiethoxysilane, silane, diethoxydiphenyl, diethoxy diphenyl silane, benzene, 1,1'-diethoxysilylene bis, acmc-1cmgu, diethoxy-di phenyl silane, diethoxydiphenylsilane, zznqqqwfkktosd-uhfffaoysa
PubChem CID 75705
IUPAC Name diethoxy(diphenyl)silane
SMILES CCO[Si](OCC)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Boiling Point 166°C to 168°C (15mmHg)
Quantity 50 g
Flash Point 175°C (347°F)
Refractive Index 1.5255
Sensitivity Moisture sensitive
Density 1.033
Beilstein 2281302
Formula Weight 272.42
Percent Purity 98%
Chemical Name or Material Diethoxydiphenylsilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.