missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1,1-cyclobutanedicarboxylate, 95%

Product Code. 11489657
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489657 50 g 50g
11479657 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489657 Supplier Thermo Scientific Alfa Aesar Supplier No. B22140.18
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3779-29-1 | C10H16O4 | 200.23 g/mol

Diethyl 1,1-cyclobutanedicarboxylate is used as a pharmaceutical intermediate. Used as intermediate in the production of Carboplatin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl 1,1-cyclobutanedicarboxylate is used as a pharmaceutical intermediate. Used as intermediate in the production of Carboplatin.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3779-29-1
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.23
MDL Number MFCD00019261
InChI Key JPNJEJSZSMXWSV-UHFFFAOYSA-N
Synonym diethyl 1,1-cyclobutanedicarboxylate, 1,1-cyclobutanedicarboxylic acid, diethyl ester, 1,1-diethyl cyclobutane-1,1-dicarboxylate, cyclobutane-1,1-dicarboxylic acid diethyl ester, 1,1-cyclobutanedicarboxylic acid diethyl ester, 1,1-cyclobutanedicarboxylic acid, 1,1-diethyl ester, 1,1-dicarboethoxycyclobutane, ethyl 1-ethoxycarbonyl cyclobutanecarboxylate, acmc-1cmcx, diethyl1,1-cyclobutanedicarboxylate
PubChem CID 77410
IUPAC Name diethyl cyclobutane-1,1-dicarboxylate
SMILES CCOC(=O)C1(CCC1)C(=O)OCC

Specifications

Density 1.05
Boiling Point 96°C to 98°C (8 mmHg)
Flash Point 89°C (192°F)
Refractive Index 1.436
Quantity 50 g
Beilstein 2050195
Solubility Information Insoluble in water.
Formula Weight 200.24
Percent Purity 95%
Chemical Name or Material Diethyl 1,1-cyclobutanedicarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.