missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1,1-cyclopropanedicarboxylate, 97%, Thermo Scientific™

Product Code. 10144130 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10144130 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10144130 Supplier Thermo Scientific Supplier No. 164450250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1559-02-0
Color Colorless
Density 1.0500g/mL
Boiling Point 94°C to 96°C (10.0 mmHg)
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C9H14O4
Linear Formula C3H4(CO2C2H5)2
Refractive Index 1.4320 to 1.434
MDL Number MFCD00001283
Quantity 25 g
Specific Gravity 1.05
Synonym diethyl 1,1-cyclopropanedicarboxylate, 1,1-diethyl cyclopropane-1,1-dicarboxylate, 1,1-cyclopropanedicarboxylic acid diethyl ester, 1,1-cyclopropanedicarboxylic acid, diethyl ester, cyclopropane-1,1-dicarboxylic acid diethyl ester, 1,1-bis ethoxycarbonyl cyclopropane, diethylcyclopropan-1,1-dicarboxylat, diethyl-1,1-cyclopropanedicarboxylate, acmc-209dcw, ksc495e2n
InChI Key KYYUCZOHNYSLFV-UHFFFAOYSA-N
SMILES CCOC(=O)C1(CC1)C(=O)OCC
IUPAC Name diethyl cyclopropane-1,1-dicarboxylate
Molecular Weight (g/mol) 186.21
PubChem CID 73790
Formula Weight 186.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diethyl 1, 1-cyclopropanedicarboxylate, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.