missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1,1-Cyclopropanedicarboxylate, TRC

Product Code. 30373291
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30373291 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373291 Supplier Toronto Research Chemicals Supplier No. TRCD44417510G
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl 1,1-Cyclopropanedicarboxylate
CAS 1559-02-0
Molecular Formula C9H14O4
Quantity 10 g
Synonym 1,1-Cyclopropanedicarboxylic Acid, 1,1-Diethyl Ester, 1,1-Bis(ethoxycarbonyl)cyclopropane, 1,1-Dicarbethoxycyclopropane, Diethyl 1,1-Cyclopropanedicarboxylate, Ethyl 1,1-Cyclopropanedicarboxylate, NSC 21376
InChI Formula InChI=1S/C9H14O4/c1-3-12-7(10)9(5-6-9)8(11)13-4-2/h3-6H2,1-2H3
SMILES CCOC(=O)C1(CC1)C(=O)OCC
Molecular Weight (g/mol) 186.21
Formula Weight 186.0892
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.