missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1,3-acetonedicarboxylate, 95%

Product Code. 10636301 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
250mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10636301 250 mL Glass bottle 250mL
10309420 50 mL Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10636301 Supplier Thermo Scientific Acros Supplier No. 325632500
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 105-50-0 | C9H14O5 | 202.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-50-0
Molecular Formula C9H14O5
Molecular Weight (g/mol) 202.21
MDL Number MFCD00009200
InChI Key ZSANYRMTSBBUCA-UHFFFAOYSA-N
Synonym diethyl 1,3-acetonedicarboxylate, diethyl 3-oxoglutarate, 1,5-diethyl 3-oxopentanedioate, pentanedioic acid, 3-oxo-, diethyl ester, ethyl acetonedicarboxylate, ethyl 3-oxoglutarate, diethyl acetone-1,3-dicarboxylate, diethyl acetone dicarboxylate, 3-oxopentanedioic acid diethyl ester, acetonedicarboxylic acid diethyl ester
PubChem CID 66045
IUPAC Name diethyl 3-oxopentanedioate
SMILES CCOC(=O)CC(=O)CC(=O)OCC

Specifications

Color Colorless to Yellow
Density 1.1120g/mL
Boiling Point 250.0°C
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C2H5O2CCH2COCH2CO2C2H5
Refractive Index 1.4380 to 1.4420
Quantity 250 mL
Beilstein 03, 791
Specific Gravity 1.112
Solubility Information Solubility in water: 10g/L in water (20°C)
Formula Weight 202.21
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Diethyl 1, 3-acetonedicarboxylate, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.