missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1-decylphosphonate, 97%

Product Code. 11327856
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327856 25 g 25g
11317856 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327856 Supplier Thermo Scientific Alfa Aesar Supplier No. L13038.14
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16165-68-7 | C14H31O3P | 278.373 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16165-68-7
Molecular Formula C14H31O3P
Molecular Weight (g/mol) 278.373
MDL Number MFCD00015593
InChI Key HYQXIVAZQBYBAF-UHFFFAOYSA-N
Synonym diethyl 1-decylphosphonate, diethyl decylphosphonate, decylphosphonic acid diethyl ester, decyldiethoxyphosphino-1-one, acmc-20al9l, diethyl n-decanephosphonate, diethyl 1-decanephosphonate, diethyl 1-decyl phosphonate, decyl-phosphonic acid diethyl ester, n-decylphosphonic acid diethyl ester
PubChem CID 2773697
IUPAC Name 1-diethoxyphosphoryldecane
SMILES CCCCCCCCCCP(=O)(OCC)OCC

Specifications

Boiling Point 128°C to 130°C (0.2 mmHg)
Refractive Index 1.439
Quantity 25 g
Beilstein 1784261
Formula Weight 278.38
Percent Purity 97%
Chemical Name or Material Diethyl 1-decylphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.