missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1-Methylbutylmalonate, TRC

Product Code. 30371363
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30371363 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30371363 Supplier Toronto Research Chemicals Supplier No. TRCD4445005G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl 1-Methylbutylmalonate
CAS 117-47-5
Molecular Formula C12H22O4
Quantity 5 g
Synonym 1-(Methylbutyl)malonic Acid Diethyl Ester, Diethyl (1-Methylbutyl)malonate, NSC 10820, NSC 62700, (1-Methylbutyl)-propanedioic Acid Diethyl Ester
InChI Formula InChI=1S/C12H22O4/c1-5-8-9(4)10(11(13)15-6-2)12(14)16-7-3/h9-10H,5-8H2,1-4H3
SMILES CCCC(C)C(C(=O)OCC)C(=O)OCC
IUPAC Name diethyl 2-pentan-2-ylpropanedioate
Molecular Weight (g/mol) 230.3
Formula Weight 230.1518
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.