missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1-tetradecylphosphonate, 97+%

Product Code. 11489397
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489397 25 g 25g
11479397 5 g 5g
11499397 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489397 Supplier Thermo Scientific Alfa Aesar Supplier No. B21426.14
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 5191-09-3 | C8H8N2 | 132.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5191-09-3
MDL Number MFCD00015597
InChI Key CPWIYYOBPHXJIA-UHFFFAOYSA-N
PubChem CID 640154
SMILES CCCCCCCCCCCCCCP(=O)(OCC)OCC

Specifications

Melting Point 8°C
Density 0.922
Boiling Point 199°C to 201°C (3 mmHg)
Molecular Formula C18H39O3P
Linear Formula CH3(CH2)13P(O)(OCH2CH3)2
Refractive Index 1.4445
Quantity 25 g
Synonym diethyl 1-tetradecanephosphonate, diethyl 1-tetradecylphosphonate, diethyl tetradecylphosphonate, acmc-20al9h, diethyl tetradecane-1-phosphonate, tetradecylphosphonic acid diethyl ester, tetradecyl-phosphonic acid diethyl ester, 1-tetradecanephosphonic acid diethyl ester, phosphonic acid, tetradecyl-, diethyl ester
IUPAC Name 1-diethoxyphosphoryltetradecane
Molecular Weight (g/mol) 334.48
Formula Weight 334.47
Percent Purity 98%
Chemical Name or Material Diethyl 1-tetradecylphosphonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.