missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2,2-diallylmalonate, Tech., Thermo Scientific™

Product Code. 10683413
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10683413 10 g 10g
10243662 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10683413 Supplier Thermo Scientific Maybridge Supplier No. SB00826EA
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 3195-24-2
Molecular Formula C13H20O4
Molecular Weight (g/mol) 240.299
MDL Number MFCD00009126
InChI Key LYUUVYQGUMRKOV-UHFFFAOYSA-N
Synonym diethyl diallylmalonate, diethyl 2,2-diallylmalonate, diallylmalonic acid diethyl ester, propanedioic acid, di-2-propenyl-, diethyl ester, 1,3-diethyl 2,2-bis prop-2-en-1-yl propanedioate, diethyl 2,2-diprop-2-enylpropane-1,3-dioate, diethyldiallylmalonate, malonic acid, diallyl-, diethyl ester, diethyl2,2-diallylmalonate, acmc-1cp69
PubChem CID 76664
IUPAC Name diethyl 2,2-bis(prop-2-enyl)propanedioate
SMILES CCOC(=O)C(CC=C)(CC=C)C(=O)OCC

Specifications

Density 0.9940g/mL
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 240.3
Percent Purity Tech.
Grade Technical
Chemical Name or Material Diethyl 2, 2-diallylmalonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.