missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2,3-Bis(benzyloxy) Tartrate, TRC

Product Code. 30379540
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30379540 100 mg 100mg
30371878 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30379540 Supplier Toronto Research Chemicals Supplier No. TRCD444855100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl 2,3-Bis(benzyloxy) Tartrate
CAS 77312-71-1
Molecular Formula C22H26O6
Quantity 100 mg
Synonym (2R,3R)-2,3-Bis(phenylmethoxy)butanedioic Acid Diethyl Ester, (2R,3R)-2,3-Bis(phenylmethoxy)butanedioic Acid 1,4-Diethyl Ester
InChI Formula InChI=1S/C22H26O6/c1-3-25-21(23)19(27-15-17-11-7-5-8-12-17)20(22(24)26-4-2)28-16-18-13-9-6-10-14-18/h5-14,19-20H,3-4,15-16H2,1-2H3/t19-,20-/m1/s1
SMILES O=C([C@H](OCC1=CC=CC=C1)[C@@H](OCC2=CC=CC=C2)C(OCC)=O)OCC
IUPAC Name diethyl (2R,3R)-2,3-bis(benzyloxy)succinate
Molecular Weight (g/mol) 386.44
Formula Weight 386.17
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.