missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (2-aminoethyl)phosphonate oxalate, 97%

Product Code. 10621952 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10621952 1 g Glass bottle 1g
10135683 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10621952 Supplier Thermo Scientific Acros Supplier No. 332220010
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 203192-98-7 | C6H17NO3P | 182.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 203192-98-7
Molecular Formula C6H17NO3P
Molecular Weight (g/mol) 182.18
MDL Number MFCD00800256
InChI Key JUAGHBASZWRMQH-UHFFFAOYSA-O
Synonym diethyl 2-aminoethyl phosphonate oxalate, diethyl aminoethyl phosphonate oxalate, 2-diethoxyphosphoryl ethylamine oxalate, 2-diethoxyphosphorylethanamine; oxalic acid, oxalic acid diethyl 2-aminoethylphosphonate, diethyl 2-aminoethyl phosphonate oxalate salt, diethyl 2-aminoethyl phosphonate ethanedioate, aminoethylphosphonic acid diethylester oxalate salt
PubChem CID 2733595
IUPAC Name 2-diethoxyphosphorylethanamine;oxalic acid
SMILES CCOP(=O)(CC[NH3+])OCC

Specifications

Melting Point 135.0°C to 141.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 271.21
Percent Purity 97%
Physical Form Crystals or Powder
Chemical Name or Material Diethyl (2-aminoethyl)phosphonate oxalate
compliance-icons
Product Identifier
  • Diethyl (2-aminoethyl)phosphonate oxalate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.