missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2-bromoethylphosphonate, 97%

Product Code. 10244380 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10244380 25 g Glass bottle 25g
10132620 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10244380 Supplier Thermo Scientific Acros Supplier No. 114180250
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5324-30-1 | C6H14BrO3P | 245.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5324-30-1
Molecular Formula C6H14BrO3P
Molecular Weight (g/mol) 245.05
MDL Number MFCD00000239
InChI Key PINITSMLVXAASM-UHFFFAOYSA-N
Synonym diethyl 2-bromoethyl phosphonate, diethyl 2-bromoethylphosphonate, phosphonic acid, 2-bromoethyl-, diethyl ester, 2-bromoethyl phosphonic acid diethyl ester, o,o-diethyl 2-bromoethyl phosphonate, diethyl bromoethylphosphonate, diethyl-2-bromoethyl-phosphonate, pinitsmlvxaasm-uhfffaoysa, diethyl-2-bromoethyl phosphonate
PubChem CID 79218
SMILES CCOP(=O)(CCBr)OCC

Specifications

Color Colorless to Yellow
Density 1.2700g/mL
Boiling Point 75°C (1.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C2H5O)2P(O)CH2CH2Br
Refractive Index 1.4590 to 1.461
Quantity 25 g
Specific Gravity 1.27
IUPAC Name 1-bromo-2-diethoxyphosphorylethane
Formula Weight 245.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diethyl 2-bromoethylphosphonate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl 2-bromoethylphosphonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.