missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2-n-Butylmalonate, TRC

Product Code. 30375256
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30375256 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30375256 Supplier Toronto Research Chemicals Supplier No. TRCD4438801G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl 2-n-Butylmalonate
CAS 133-08-4
Molecular Formula C11H20O4
Quantity 1 g
Synonym 2-butylpropanedioic Acid 1,3-Diethyl Ester, Butylmalonic Acid Diethyl Ester, Butylpropanedioic Acid Diethyl Ester, Diethyl 2-Butylmalonate, Diethyl Butylmalonate, Diethyl n-Butylmalonate, Diethyl alpha-Butylmalonate, Ethyl Butylmalonate, NSC 4565
InChI Formula InChI=1S/C11H20O4/c1-4-7-8-9(10(12)14-5-2)11(13)15-6-3/h9H,4-8H2,1-3H3
SMILES CCCCC(C(=O)OCC)C(=O)OCC
IUPAC Name diethyl 2-butylpropanedioate
Molecular Weight (g/mol) 216.27
Formula Weight 216.1362
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.