missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (3-aminopropyl)phosphonate, 95%, Thermo Scientific Chemicals

Product Code. 15488367
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488367 250 mg 250mg
15478367 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488367 Supplier Thermo Scientific Alfa Aesar Supplier No. H51874.MD
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4402-24-8 | Molecular Formula: C7H18NO3P | Molecular Weight: 195.20 g/mol

Diethyl (3-aminopropyl)phosphonate is used in the preparation of tert-Butyl (2-((diethoxyphosphoryl)propyl)amino)-3,4-dioxocyclobut-1- en-1-yl)carbamate by reacting with tert-butyl (2-ethoxy-3,4-dioxocyclobut-1-en-1-yl)carbamate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl (3-aminopropyl)phosphonate is used in the preparation of tert-Butyl (2-((diethoxyphosphoryl)propyl)amino)-3,4-dioxocyclobut-1- en-1-yl)carbamate by reacting with tert-butyl (2-ethoxy-3,4-dioxocyclobut-1-en-1-yl)carbamate.

Notes
Air sensitive. Incompatible with oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4402-24-8
Molecular Formula C7H18NO3P
Molecular Weight (g/mol) 195.20
MDL Number MFCD11110596
InChI Key DQORFBNFNLHVIM-UHFFFAOYSA-N
Synonym diethyl 3-aminopropyl phosphonate, diethyl 3-aminopropylphosphonate, 3-aminopropyl phosphonic acid diethyl ester, diethyl aminopropyl phosphonate, 3-aminopropylphosphonic acid diethyl ester, 3-aminopropanephosphonic acid diethyl ester, 3-amino-propyl-phosphonic acid diethyl ester, diethyl 3-aminopropyl phosphonate 250mg, phosphonic acid, 3-aminopropyl-, diethyl ester
PubChem CID 78113
IUPAC Name 3-diethoxyphosphorylpropan-1-amine
SMILES CCOP(=O)(CCCN)OCC

Specifications

Quantity 250 mg
UN Number UN2735
Formula Weight 195.2
Percent Purity 95%
Chemical Name or Material Diethyl (3-aminopropyl)phosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.