missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 4-methylbenzylphosphonate, 98+%

Product Code. 11422208
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11422208 50 g 50g
11412208 10 g 10g
11432208 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11422208 Supplier Thermo Scientific Alfa Aesar Supplier No. A15190.18
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3762-25-2 | C12H19O3P | 242.255 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3762-25-2
Molecular Formula C12H19O3P
Molecular Weight (g/mol) 242.255
MDL Number MFCD00041385
InChI Key QKGBKPZAXXBLJE-UHFFFAOYSA-N
Synonym diethyl 4-methylbenzylphosphonate, diethyl 4-methylbenzyl phosphonate, 4-methylbenzyl phosphonic acid diethyl ester, diethyl 4-methylphenyl methylphosphonate, diethoxy 4-methylphenyl methyl phosphino-1-one, acmc-209iuf, diethyl-4-methylbenzylphosphonate, 1-diethoxyphosphorylmethyl-4-methylbenzene, phosphonic acid, 4-methylphenyl methyl-, diethyl ester
PubChem CID 3050
IUPAC Name 1-(diethoxyphosphorylmethyl)-4-methylbenzene
SMILES CCOP(=O)(CC1=CC=C(C=C1)C)OCC

Specifications

Density 1.07
Boiling Point 146°C to 148°C (0.6 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.496
Quantity 50 g
Beilstein 2940626
Formula Weight 242.26
Percent Purity ≥98%
Chemical Name or Material Diethyl 4-methylbenzylphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.