missing translation for 'onlineSavingsMsg'
Learn More

Diethyl acetylmethylphosphonate, 97%, Thermo Scientific Chemicals

Product Code. 11481440
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11481440 5 g 5g
11471440 1 g 1g
11491440 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11481440

Brand: Thermo Scientific Alfa Aesar H26896.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Diethyl acetylmethylphosphonate is used as ligand in coordination chemistry to prepare lanthanide series metals like terbium, europium and neodymium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl acetylmethylphosphonate is used as ligand in coordination chemistry to prepare lanthanide series metals like terbium, europium and neodymium.

Solubility
Miscible with tetrahydrofuran, ether, dichloromethane and chloroform.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1067-71-6
Molecular Formula C7H15O4P
Molecular Weight (g/mol) 194.167
MDL Number MFCD00044728
InChI Key RSAFKRSMGOSHRK-UHFFFAOYSA-N
Synonym diethyl 2-oxopropyl phosphonate, diethyl acetonylphosphonate, diethyl 2-oxopropylphosphonate, acetonylphosphonic acid diethyl ester, 2-oxopropyl phosphonic acid diethyl ester, phosphonic acid, 2-oxopropyl-, diethyl ester, aurora ka-1476, ksc490a4t, diethyl acetylmethylphosphonate
PubChem CID 14028
IUPAC Name 1-diethoxyphosphorylpropan-2-one
SMILES CCOP(=O)(CC(=O)C)OCC

Specifications

Density 1.01
Boiling Point 129°C (9 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.433
Quantity 5 g
Solubility Information Miscible with tetrahydrofuran,ether,dichloromethane and chloroform.
Formula Weight 194.17
Percent Purity 97%
Chemical Name or Material Diethyl acetylmethylphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.