missing translation for 'onlineSavingsMsg'
Learn More

Diethyl acetylmethylphosphonate, 97%, Thermo Scientific Chemicals

Product Code. 11491440
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491440 25 g 25g
11471440 1 g 1g
11481440 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491440 Supplier Thermo Scientific Alfa Aesar Supplier No. H26896.14
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Diethyl acetylmethylphosphonate is used as ligand in coordination chemistry to prepare lanthanide series metals like terbium, europium and neodymium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl acetylmethylphosphonate is used as ligand in coordination chemistry to prepare lanthanide series metals like terbium, europium and neodymium.

Solubility
Miscible with tetrahydrofuran, ether, dichloromethane and chloroform.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1067-71-6
Molecular Formula C7H15O4P
Molecular Weight (g/mol) 194.167
MDL Number MFCD00044728
InChI Key RSAFKRSMGOSHRK-UHFFFAOYSA-N
Synonym diethyl 2-oxopropyl phosphonate, diethyl acetonylphosphonate, diethyl 2-oxopropylphosphonate, acetonylphosphonic acid diethyl ester, 2-oxopropyl phosphonic acid diethyl ester, phosphonic acid, 2-oxopropyl-, diethyl ester, aurora ka-1476, ksc490a4t, diethyl acetylmethylphosphonate
PubChem CID 14028
IUPAC Name 1-diethoxyphosphorylpropan-2-one
SMILES CCOP(=O)(CC(=O)C)OCC

Specifications

Density 1.01
Boiling Point 129°C (9 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.433
Quantity 25 g
Solubility Information Miscible with tetrahydrofuran,ether,dichloromethane and chloroform.
Formula Weight 194.17
Percent Purity 97%
Chemical Name or Material Diethyl acetylmethylphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.