missing translation for 'onlineSavingsMsg'
Learn More

Diethyl acetylsuccinate, 97+%

Product Code. 11490168
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490168 5 g 5g
11400178 25 g 25g
11410178 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490168 Supplier Thermo Scientific Alfa Aesar Supplier No. B23744.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1115-30-6 | C10H16O5 | 216.233 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1115-30-6
Molecular Formula C10H16O5
Molecular Weight (g/mol) 216.233
MDL Number MFCD00009157
InChI Key DVSDDICSXBCMQJ-UHFFFAOYSA-N
Synonym diethyl 2-acetylsuccinate, diethyl acetylsuccinate, ethyl acetylsuccinate, diethyl acetosuccinate, butanedioic acid, acetyl-, diethyl ester, acetylsuccinic acid diethyl ester, diethylacetylsuccinate, 1,4-diethyl 2-acetylbutanedioate, succinic acid, acetyl-, diethyl ester, butanedioic acid, 2-acetyl-, 1,4-diethyl ester
PubChem CID 66197
IUPAC Name diethyl 2-acetylbutanedioate
SMILES CCOC(=O)CC(C(=O)C)C(=O)OCC

Specifications

Melting Point -8°C
Density 1.081
Boiling Point 180°C to 183°C (50 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4346
Quantity 5 g
Formula Weight 216.23
Percent Purity ≥97%
Chemical Name or Material Diethyl acetylsuccinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.