missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Adipate, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Diethyl adipate |
| CAS | 141-28-6 |
| Molecular Formula | C10 H18 O4 |
| Quantity | 25 g |
| Synonym | Adipic acid, diethyl ester (6CI,8CI), Hexanedioic acid, diethyl ester (9CI), Diethyl adipate, Diethyl hexanedioate, Ethyl adipate, Ethyl δ-carboethoxyvalerate, NSC 19160, NSC 3363 |
| InChI Formula | InChI=1S/C10H18O4/c1-3-13-9(11)7-5-6-8-10(12)14-4-2/h3-8H2,1-2H3 |
| SMILES | CCOC(=O)CCCCC(=O)OCC |
| IUPAC Name | diethyl hexanedioate |
| Molecular Weight (g/mol) | 202.25 |
| Formula Weight | 202.1205 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction