missing translation for 'onlineSavingsMsg'
Learn More

Diethyl allyl(3-phenyl-2-propynyl)malonate, 96%, Thermo Scientific Chemicals

Product Code. 11342809
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342809 1 g 1g
11352809 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342809 Supplier Thermo Scientific Alfa Aesar Supplier No. H27702.03
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 143633-91-4 | Molecular Formula: C19H22O4 | Molecular Weight: 314.381 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143633-91-4
Molecular Formula C19H22O4
Molecular Weight (g/mol) 314.381
MDL Number MFCD09953493
InChI Key ZSPBIVGRCZUELN-UHFFFAOYSA-N
Synonym diethyl allyl 3-phenyl-2-propynyl malonate, diethyl 2-allyl-2-3-phenylprop-2-yn-1-yl malonate, propanedioic acid,2-3-phenyl-2-propyn-1-yl-2-2-propen-1-yl-, 1,3-diethyl ester, 1,3-diethyl 2-3-phenylprop-2-yn-1-yl-2-prop-2-en-1-yl propanedioate, acmc-20ed36, 3-phenylpropargylallylmalonic acid diethyl ester, allyl 3-phenyl-2-propynyl malonic acid diethyl ester, diethyl 1-phenyl-6-heptene-1-yne-4,4-dicarboxylate, diethyl 3-phenylprop-2-yn-1-yl prop-2-en-1-yl propanedioate
PubChem CID 10086992
IUPAC Name diethyl 2-(3-phenylprop-2-ynyl)-2-prop-2-enylpropanedioate
SMILES CCOC(=O)C(CC=C)(CC#CC1=CC=CC=C1)C(=O)OCC

Specifications

Refractive Index 1.514
Quantity 1 g
Formula Weight 314.38
Percent Purity 96%
Chemical Name or Material Diethyl allyl(3-phenyl-2-propynyl)malonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.