missing translation for 'onlineSavingsMsg'
Learn More

Diethyl allylmalonate, 97%

Product Code. 10050890 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10050890

Brand: Thermo Scientific Acros 113961000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 2049-80-1 | C10H16O4 | 200.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2049-80-1
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.23
MDL Number MFCD00009155
InChI Key GDWAYKGILJJNBB-UHFFFAOYSA-N
Synonym diethyl allylmalonate, ethyl allylmalonate, diethyl 2-allylmalonate, allylmalonic acid diethyl ester, diethyl prop-2-enylpropanedioate, diethyl 2-prop-2-enyl malonate, malonic acid, allyl-, diethyl ester, propanedioic acid, 2-propenyl-, diethyl ester, diethyl 2-2-propenyl-1,3-propanedioate, allyl malonic acid
PubChem CID 74900
IUPAC Name diethyl 2-prop-2-enylpropanedioate
SMILES CCOC(=O)C(CC=C)C(=O)OCC

Specifications

Color Colorless to Yellow
Density 1.0100g/mL
Boiling Point 222.0°C to 223.0°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C2H5O2CCH(CH2CH=CH2)CO2C2H5
Refractive Index 1.4300 to 1.4320
Quantity 100 g
Beilstein 02, 776
Specific Gravity 1.01
Formula Weight 200.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diethyl allylmalonate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl allylmalonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.