missing translation for 'onlineSavingsMsg'
Learn More

Diethyl azodicarboxylate, 97%, Thermo Scientific™

Product Code. 11394266
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394266 100 g 100g
11374266 5 g 5g
11384266 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11394266 Supplier Thermo Scientific Alfa Aesar Supplier No. L19348.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Diethyl azodicarboxylate acts as a dienophile used in Diels-Alder reactions. It is used as a reagent in alfa-thiocyanation of enolizable ketones with ammonium thiocyanate and annulation of N-protected imines. It acts as a reactant for the preparation of immunostimulants alfa-Galactosylceramides, bisubstrate inhibitors with molecular recognition at the active site of catechol-O-methyltransferase and aza-beta-lactams via NHC-catalyzed [2 + 2] cycloaddition with ketenes. It finds application in Mitsunobu reaction and used in the synthesis of pharmaceuticals like zidovudine and procarbazine. The presence of azo group in it is a Michael acceptor and used for the conversion of beta-ketoesters to the corresponding hydrazine derivatives in presence of copper(II) catalyst. Further, it is an efficient dehydrogenating agent, which is involved in the preparation of aldehydes, disulfides from alcohols and thiols respectively.

Solubility
Miscible with dichloromethane, diethyl ether and toluene.

Notes
Store in a cool place. Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1972-28-7
Molecular Formula C6H10N2O4
Molecular Weight (g/mol) 174.156
MDL Number MFCD00009103
InChI Key FAMRKDQNMBBFBR-BQYQJAHWSA-N
Synonym diethyl azodicarboxylate, dead, diethyl azodiformate, ethyl azodicarboxylate, diethoxycarbonyldiazene, azodicarboxylic acid diethyl ester, diethyl e-diazene-1,2-dicarboxylate, ethyl ne-n-ethoxycarbonyliminocarbamate, diethyl diazenedicarboxylate, diazenedicarboxylic acid, diethyl ester
PubChem CID 5462977
IUPAC Name ethyl (NE)-N-ethoxycarbonyliminocarbamate
SMILES CCOC(=O)N=NC(=O)OCC

Specifications

Density 1.106
Boiling Point 106°C (13 mmHg)
Flash Point 85°C (185°F)
Linear Formula CH3CH2O2C-N=N-CO2CH2CH3
Refractive Index 1.421
Quantity 100 g
UN Number UN2810
Beilstein 908662
Sensitivity Air and light sensitive
Solubility Information Miscible with dichloromethane,diethyl ether and toluene.
Formula Weight 174.16
Percent Purity 97%
Chemical Name or Material Diethyl azodicarboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.