missing translation for 'onlineSavingsMsg'
Learn More

Diethyl benzylmalonate, 97%

Product Code. 11499937
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499937 100 g 100g
11489937 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499937 Supplier Thermo Scientific Alfa Aesar Supplier No. B22890.22
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 607-81-8 | C14H18O4 | 250.294 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 607-81-8
Molecular Formula C14H18O4
Molecular Weight (g/mol) 250.294
MDL Number MFCD00009166
InChI Key ICZLTZWATFXDLP-UHFFFAOYSA-N
Synonym diethyl benzylmalonate, diethyl 2-benzylmalonate, benzylmalonic acid diethyl ester, propanedioic acid, phenylmethyl-, diethyl ester, unii-40y0x0xn6p, malonic acid, benzyl-, diethyl ester, malonic acid, 2-benzyl-, diethyl ester, diethyl benzyl malonate, 1,3-diethyl 2-benzylpropanedioate, diethyl benzylpropanedioate
PubChem CID 69090
IUPAC Name diethyl 2-benzylpropanedioate
SMILES CCOC(=O)C(CC1=CC=CC=C1)C(=O)OCC

Specifications

Density 1.069
Boiling Point 162°C to 163°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.486
Quantity 100 g
Beilstein 615793
Formula Weight 250.29
Percent Purity 97%
Chemical Name or Material Diethyl benzylmalonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.