missing translation for 'onlineSavingsMsg'
Learn More

Diethyl benzylphosphonate, 99%

Product Code. 11965141 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11965141 500 g Glass bottle 500g
10162570 25 g Glass bottle 25g
10122620 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11965141 Supplier Thermo Scientific Acros Supplier No. 114175000
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1080-32-6 | C11H17O3P | 228.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1080-32-6
Molecular Formula C11H17O3P
Molecular Weight (g/mol) 228.23
InChI Key AIPRAPZUGUTQKX-UHFFFAOYSA-N
Synonym diethyl benzylphosphonate, benzylphosphonic acid diethyl ester, diethoxyphosphonomethylbenzene, diethyl phosphonate, benzyl, phosphonic acid, phenylmethyl-, diethyl ester, unii-t6d4qwt656, diethylbenzylphosphonate, phosphonic acid, benzyl-, diethyl ester, diethyl phenylmethyl phosphonate, phosphonic acid, p-phenylmethyl-, diethyl ester
PubChem CID 14122
IUPAC Name diethoxyphosphorylmethylbenzene
SMILES CCOP(=O)(CC1=CC=CC=C1)OCC

Specifications

Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 106.0°C to 108.0°C (1.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (C2H5O)2P(O)CH2C6H5
Refractive Index 1.4950 to 1.4980
Quantity 500 g
Specific Gravity 1.09
Formula Weight 228.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diethyl benzylphosphonate
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl benzylphosphonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.