missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Bromodifluoromethylphosphonate, TRC

Product Code. 30388347
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30388347 10 g 10g
30384462 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30388347 Supplier Toronto Research Chemicals Supplier No. TRCD44384510G
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl Bromodifluoromethylphosphonate
CAS 65094-22-6
Molecular Formula C5H10BrF2O3P
Quantity 10 g
Synonym P-(Bromodifluoromethyl)phosphonic Acid Diethyl Ester, (Bromodifluoromethyl)phosphonic Acid Diethyl Ester
InChI Formula InChI=1S/C5H10BrF2O3P/c1-3-10-12(9,11-4-2)5(6,7)8/h3-4H2,1-2H3
SMILES CCOP(=O)(OCC)C(F)(F)Br
IUPAC Name 1-[[bromo(difluoro)methyl]-ethoxyphosphoryl]oxyethane
Molecular Weight (g/mol) 267.01
Formula Weight 265.9519
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.