missing translation for 'onlineSavingsMsg'
Learn More

Diethyl carbonate, 99%

Product Code. 11494084
Change view
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
Unit Size:
10000mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494084 2500 mL 2500mL
11484084 500 mL 500mL
11404094 10,000 mL 10000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494084 Supplier Thermo Scientific Alfa Aesar Supplier No. A12477.0F
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 105-58-8 | C5H10O3 | 118.13 g/mol

Diethyl carbonate is esters, beta-enamino esters, carbamates and unsymmetrical alkyl carbonates. It is an active component of electrolytes used in lithium which is used as a solvent for cellulose ethers, nitro cellulose, natural and synthetic resin and in erythromycin intramuscular injections. It is also used in the synthesis of 3-ethyl-4-methyl-5-phenyl-3H-oxazol-2-one, phenobarbital, pyrethrum batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl carbonate is esters, beta-enamino esters, carbamates and unsymmetrical alkyl carbonates. It is an active component of electrolytes used in lithium which is used as a solvent for cellulose ethers, nitro cellulose, natural and synthetic resin and in erythromycin intramuscular injections. It is also used in the synthesis of 3-ethyl-4-methyl-5-phenyl-3H-oxazol-2-one, phenobarbital, pyrethrum batteries.

Solubility
Miscible with diehtyl ether, ethanol and chloroform.Immiscible with water.

Notes
Store in cool place. Moisture sensitive. Incompatible with strong acids, oxidizing agents, strong bases and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-58-8
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00009107
InChI Key OIFBSDVPJOWBCH-UHFFFAOYSA-N
Synonym ethyl carbonate, carbonic acid, diethyl ester, carbonic acid diethyl ester, eufin, diatol, diaethylcarbonat, carbonic ether, ethoxyformic anhydride, diethylkarbonat, ethyl carbonate eto 2co
PubChem CID 7766
IUPAC Name diethyl carbonate
SMILES CCOC(=O)OCC

Specifications

Melting Point -43°C
Density 0.975
Boiling Point 126°C to 128°C
Flash Point 31°C (87°F)
Assay Percent Range ≥99%
Odor Pleasant
Refractive Index 1.3845
Quantity 2500 mL
UN Number UN2366
Beilstein 956591
Sensitivity Moisture sensitive
Merck Index 14,3780
Solubility Information Miscible with diehtyl ether,ethanol and chloroform.Immiscible with water.
Formula Weight 118.13
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Diethyl carbonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.