missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Carbonate, TRC

Product Code. 30387348
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30387348 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387348 Supplier Toronto Research Chemicals Supplier No. TRCD44400010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl Carbonate
CAS 105-58-8
Purity Grade Notes GC
Molecular Formula C5H10O3
Quantity 10 g
Synonym DEC, Diatol, Diatol (Carbonate), Diethyl Carbonate, Diethyl Ester Carbonic Acid, Ethyl Carbonate, Ethyl Carbonate ((EtO)2CO), Eufin, H-DEC, NSC 8849
InChI Formula InChI=1S/C5H10O3/c1-3-7-5(6)8-4-2/h3-4H2,1-2H3
SMILES CCOC(=O)OCC
Molecular Weight (g/mol) 118.03
Formula Weight 118.063
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.