missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Chloromalonate, TRC

Product Code. 30385165
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385165 10 g
30381890 1 g
30372592 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385165 Supplier Toronto Research Chemicals Supplier No. TRCD44406010G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14064-10-9
Molecular Formula C7H11ClO4
Molecular Weight (g/mol) 194.61
Synonym 2-Chloropropanedioic Acid 1,3-Diethyl Ester, Chloromalonic Acid Diethyl Ester, Chloropropanedioic Acid Diethyl Ester, Chloromalonic Acid Diethyl Ester, Chloropropanedioic Acid Ethyl Ester, Diethyl Monochloromalonate, Ethyl Chloromalonate, NSC 227952, alpha-Chlorodiethyl Malonate
IUPAC Name diethyl 2-chloropropanedioate
SMILES CCOC(=O)C(Cl)C(=O)OCC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C7H11ClO4/c1-3-11-6(9)5(8)7(10)12-4-2/h5H,3-4H2,1-2H3
Formula Weight 194.0346
Chemical Name or Material Diethyl Chloromalonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.