missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Chlorophosphite, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Diethyl Chlorophosphite |
| CAS | 589-57-1 |
| Molecular Formula | C4H10ClO2P |
| Quantity | 10 g |
| Synonym | Ethyl phosphorochloridite, Chlorodiethoxyphosphine, Diethoxychlorophosphine, Diethyl Chlorophosphonite, Diethyl Phosphorochloridite, Ethyl Chlorophosphite, Ethyl Phosphorochloridite, (EtO)2ClP |
| InChI Formula | InChI=1S/C4H10ClO2P/c1-3-6-8(5)7-4-2/h3-4H2,1-2H3 |
| SMILES | CCOP(Cl)OCC |
| IUPAC Name | chloro(diethoxy)phosphane |
| Molecular Weight (g/mol) | 156.55 |
| Formula Weight | 156.0107 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction