missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Chlorothiophosphate, TRC

Product Code. 30386292
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30386292 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386292 Supplier Toronto Research Chemicals Supplier No. TRCD4441601G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl phosphorochloridothionate
CAS 2524-04-1
Molecular Formula C4 H10 Cl O2 P S
Quantity 1 g
Synonym Ethyl phosphorochloridothioate ((EtO)2ClPS) (6CI), (Chlorodiethoxy)phosphine sulfide, Diethoxythiophosphoryl chloride, Diethyl chlorothiophosphate, Diethyl phosphorochloridothioate, Diethyl phosphorochlorothioate, Diethyl phosphorothiochloridate, Diethyl phosphorothionochloridate, Diethyl thiophosphoryl chloride, NSC 43776, O,O-Diethyl chloridophosphorothioate, O,O-Diethyl chloridothionophosphate, O,O-Diethyl chlorophosphorothioate, O,O-Diethyl chlorothionophosphate, O,O-Diethyl chlorothiophosphate, O,O-Diethyl chlorothiophosphonate, O,O-Diethyl phosphorochloridothioate, O,O-Diethyl phosphorochlorothioate, O,O-Diethyl phosphorothiochloridate, O,O-Diethyl phosphorothionochloridate, O,O-Diethyl thionophosphoric chloride, O,O-Diethyl thionophosphorochloridate, O,O-Diethyl thionophosphoryl chloride, O,O-Diethyl thiophosphoric acid chloride, O,O-Diethyl thiophosphorochloridate, O,O-Diethyl thiophosphoryl chloride
InChI Formula InChI=1S/C4H10ClO2PS/c1-3-6-8(5,9)7-4-2/h3-4H2,1-2H3
SMILES CCOP(=S)(Cl)OCC
IUPAC Name chloro-diethoxy-sulfanylidene-λ^{5}-phosphane
Molecular Weight (g/mol) 188.61
Formula Weight 187.9828
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.