missing translation for 'onlineSavingsMsg'
Learn More

Diethyl cyclopropane-1,1-dicarboxylate

Product Code. 15493068
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493068 25 g 25g
15473068 1 g 1g
15483068 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15493068 Supplier Thermo Scientific Alfa Aesar Supplier No. H56568.14
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1559-02-0 | C9H14O4 | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1559-02-0
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.21
MDL Number MFCD00001283
InChI Key KYYUCZOHNYSLFV-UHFFFAOYSA-N
Synonym diethyl 1,1-cyclopropanedicarboxylate, 1,1-diethyl cyclopropane-1,1-dicarboxylate, 1,1-cyclopropanedicarboxylic acid diethyl ester, 1,1-cyclopropanedicarboxylic acid, diethyl ester, cyclopropane-1,1-dicarboxylic acid diethyl ester, 1,1-bis ethoxycarbonyl cyclopropane, diethylcyclopropan-1,1-dicarboxylat, diethyl-1,1-cyclopropanedicarboxylate, acmc-209dcw, ksc495e2n
PubChem CID 73790
IUPAC Name diethyl cyclopropane-1,1-dicarboxylate
SMILES CCOC(=O)C1(CC1)C(=O)OCC

Specifications

Density 1.055
Boiling Point 94°C to 96°C (10 mmHg)
Flash Point 87°C (189°F)
Odor Odorless
Refractive Index 1.433
Quantity 25 g
Beilstein 2049509
Formula Weight 186.21
Chemical Name or Material Diethyl cyclopropane-1,1-dicarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.