missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl-d10-amine HCl, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Diethylamine-D10 Hydrochloride |
| CAS | 285132-87-8 |
| Molecular Formula | C4 D10 H N . Cl H |
| Quantity | 0.5 g |
| Synonym | Ethan-d5-amine, N-(ethyl-d5)-, hydrochloride (9 CI), 1,1,2,2,2-Pentadeuterio-N-(1,1,2,2,2-pentadeuterioethyl)ethanamine hydrochloride, N-(Ethyl-d5)ethan-d5-amine hydrochloride, Diethyl-d10-amine hydrochloride, Diethylamine-d10 hydrochloride, Diethylamine-D10 hydrochloride |
| InChI Formula | InChI=1 S/C4H11N.ClH/c1-3-5-4-2;/h5H,3-4H2,1-2H3;1 H/i1D3,2D3,3D2,4D2; |
| SMILES | Cl.[2 H]C([2 H])([2 H])C([2 H])([2 H])NC([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,2,2,2-pentadeuterio-N-(1,1,2,2,2-pentadeuterioethyl)ethanamine;hydrochloride |
| Molecular Weight (g/mol) | 119.6594 |
| Formula Weight | 119.1286 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction