missing translation for 'onlineSavingsMsg'
Learn More

Diethyl dimethylaminomethylenemalonate, 94%

Product Code. 11455646
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455646 25 g 25g
11445646 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455646 Supplier Thermo Scientific Alfa Aesar Supplier No. B20765.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 18856-68-3 | C10H17NO4 | 215.25 g/mol

Derivatives of this compound, diethyl ethoxymethylenemalonate is used in the synthesis of chemotherapeutic agents using 6-aminobenzothiazoles as starting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Derivatives of this compound, diethyl ethoxymethylenemalonate is used in the synthesis of chemotherapeutic agents using 6-aminobenzothiazoles as starting material.

Solubility
Soluble in methanol.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18856-68-3
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.25
MDL Number MFCD00052394
InChI Key XNSXXTKUKYWLAH-UHFFFAOYSA-N
Synonym diethyl dimethylaminomethylenemalonate, diethyl 2-dimethylamino methylidene malonate, diethyl 2-dimethylaminomethylidene propanedioate, diethyl 2-dimethylamino methylene malonate, propanedioic acid, dimethylamino methylene-, diethyl ester, malonic acid, dimethylamino-methyl-, diethyl ester, malonic acid, dimethylaminomethylene-, diethyl ester, 1,3-diethyl 2-dimethylamino methylidene propanedioate, acmc-1buy0
PubChem CID 140439
IUPAC Name diethyl 2-(dimethylaminomethylidene)propanedioate
SMILES CCOC(=O)C(=CN(C)C)C(=O)OCC

Specifications

Boiling Point 134°C to 136°C (0.5 mmHg)
Quantity 25 g
Solubility Information Soluble in methanol.
Formula Weight 215.25
Percent Purity 95%
Chemical Name or Material Diethyl dimethylaminomethylenemalonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.