missing translation for 'onlineSavingsMsg'
Learn More

Diethyl disulfide, 99%

Product Code. p-7045264
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11332417

Brand: Thermo Scientific Alfa Aesar L10065.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-81-6 | C4H10S2 | 122.244 g/mol

Diethyl sulfide is used as a solvent for anhydrous minerals and in plating baths for gold and silver

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl sulfide is used as a solvent for anhydrous minerals and in plating baths for gold and silver

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-81-6
Molecular Formula C4H10S2
Molecular Weight (g/mol) 122.244
MDL Number MFCD00009266
InChI Key CETBSQOFQKLHHZ-UHFFFAOYSA-N
Synonym diethyl disulfide, ethyl disulfide, disulfide, diethyl, diethyl disulphide, ethyldithioethane, 3,4-dithiahexane, ethyl disulphide, ethyldisulfanyl ethane, diethyldisulfid, diethyldisulfide
PubChem CID 8077
IUPAC Name (ethyldisulfanyl)ethane
SMILES CCSSCC

Specifications

Melting Point -102°C
Density 0.993
Boiling Point 152°C to 154°C
Flash Point 40°C (104°F)
Odor Stench
Linear Formula CH3CH2S-SCH2CH3
Refractive Index 1.506
Quantity 25 g
UN Number UN1993
Beilstein 1098273
Solubility Information Soluble in water.
Formula Weight 122.25
Percent Purity 99%
Chemical Name or Material Diethyl disulfide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.