missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether, ACS Reagent, contains BHT as inhibitor, anhydrous, ≥99.0%, Honeywell Riedel-de Haën™

Product Code. 15664310
Change view
Click to view available options
Quantity:
100 mL
250 mL
1 L
6 x 1 L
Packaging:
Glass Bottle
Unit Size:
1 L
100mL
250mL
Pack of 6
Product Code. Quantity Packaging unitSize
15664310 1 L Glass Bottle 1 L
15654310 100 mL Glass Bottle 100mL
15674310 250 mL Glass Bottle 250mL
15684310 6 x 1 L Glass Bottle Pack of 6
4 options
This item is not returnable. View return policy

Product Code. 15664310

Brand: Honeywell-Riedel-de Haen 3461361L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ACS Reagent, contains BHT as inhibitor, anhydrous,≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116°C (literature)
CAS Max % ≤100.0000%
Density 0.706 g/mL at 25 °C(lit.)
Boiling Point 34°C to 35°C
Quantity 1 L
Flash Point −45°C
Packaging Glass Bottle
Physical Form anhydrous
Linear Formula (CH3CH2)2O
UN Number UN1155
Beilstein 1696894
Formula Weight 74.12
Percent Purity >99%
Grade ACS Reagent
Assay (GC)
Chemical Name or Material Diethyl ether, ACS Reagent, contains BHT as inhibitor, anhydrous, ≥99.0%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.