missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Ether, Certified AR for Analysis, Stabilised with BHT, meets analytical specification of Ph.Eur, Fisher Chemical™

Product Code. 10696442 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
Aluminum Can
Amber Glass Bottle
Metal Can
Plastic-coated Glass Bottle
Unit Size:
1 L
2.5 L
25 L
5 L
500mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10696442 500 mL Plastic-coated Glass Bottle 500mL
10306040 1 L Amber Glass Bottle 1 L
10111570 1 L Plastic-coated Glass Bottle 1 L
10785901 2.5 L Amber Glass Bottle 2.5 L
10171610 2.5 L Plastic-coated Glass Bottle 2.5 L
11984691 5 L Aluminum Can 5 L
11323440 25 L Metal Can 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 10696442 Supplier Fisher Chemical Supplier No. D/2450/PB08
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

C4H10O, CAS Number-60-29-7, ether, pronarcol, 3-oxapentane, anesthetic ether, aether, anaesthetic ether, diethyl oxide, diethyl ether, ethyl ether, ethyl oxide, 2.5L, 34.6 deg.C, CHEBI:35702, Colorless, 74.12g/mol, RTZKZFJDLAIYFH-UHFFFAOYSA-N, ethoxyethane, -116 deg.C, 11646, 74.123 | CAS: 60-29-7 | C4H10O | 74.12 g/mol

C4H10O, CAS Number-60-29-7, ether, pronarcol, 3-oxapentane, anesthetic ether, aether, anaesthetic ether, diethyl oxide, diethyl ether, ethyl ether, ethyl oxide, 2.5L, 34.6 deg.C, CHEBI:35702, Colorless, 74.12g/mol, RTZKZFJDLAIYFH-UHFFFAOYSA-N, ethoxyethane, -116 deg.C, 11646, 74.123

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116°C
Color Colorless
Quantity 500 mL
Packaging Plastic-coated Glass Bottle
Physical Form Liquid
Vapor Pressure 587mbar at 20°C
Viscosity 0.2448cp at 20°C
Boiling Point 34.6°C
Formula Weight 74.12g/mol
compliance-icons compliance-icons
Product Identifier
  • Diethyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.