missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Ether, for HPLC, Stabilised with Ethanol, Fisher Chemical™

Product Code. 10407830 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber Glass Bottle
Plastic-coated Glass Bottle
Unit Size:
1 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10407830 1 L Amber Glass Bottle 1 L
10579950 2.5 L Amber Glass Bottle 2.5 L
10050100 2.5 L Plastic-coated Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10407830 Supplier Fisher Chemical Supplier No. D/2506/15
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Identificadores de productos químicos

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Especificaciones

Melting Point -116°C
Color Colorless
Quantity 1 L
Packaging Amber Glass Bottle
Physical Form Liquid
Vapor Pressure 587mbar at 20°C
Viscosity 0.2448cp at 20°C
Boiling Point 34.6°C
Formula Weight 74.12g/mol
compliance-icons compliance-icons
Nombre del producto
  • Diethyl ether
Palabra de advertencia
  • Danger
Clasificación
  • Flammable liquid Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.