missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Ether, puriss., ≥99.5% (GC), Honeywell™ Riedel-de Haën™
BIOPHARMA

Product Code. 15662460
Change view
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
4 x 5 L
6 x 1 L
Packaging:
Aluminum Bottle
Glass Bottle
Unit Size:
2.5 L
Pack of 4
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15662460 4 x 2.5 L Glass Bottle Pack of 4
15632460 2.5 L Aluminum Bottle 2.5 L
15602470 6 x 1 L Glass Bottle Pack of 6
15672460 4 x 5 L Aluminum Bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15662460 Supplier Honeywell-Riedel-de Haen Supplier No. 240044X2.5L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur.,≥99.5% (GC)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116°C
Density 0.706g/mL at 25°C
Quantity 4 x 2.5 L
Explosion Limit 0.365
Flash Point −45°C (−49°F)
Ignition Point 320°F
Packaging Glass Bottle
Physical Form Liquid
Linear Formula (CH3CH2)2O
UN Number UN1155
Beilstein 1696894
Boiling Point 34.6°C
Formula Weight 74.12g/mol
Percent Purity ≥99.5% (GC)
Grade puriss.
Chemical Name or Material Diethyl Ether, Meets analytical specification of Ph.Eur.BP
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.