missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether, Solstice

Product Code. 15974750
Change view
Click to view available options
Quantity:
18 L
Packaging:
Returnable Stainless Steel Can
Unit Size:
18 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15974750 18 L Returnable Stainless Steel Can 18 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15974750 Supplier Honeywell-Riedel-de Haen Supplier No. 3494218L
  • ONLINE EXCLUSIVE PRICE
    2235.00 €
    2023.37 € / 18 L
    Save 211.63 €
    9% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, dried, stabilized, max.0,0075% H2O

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -116°C (1,013 hPa)
CAS 60-29-7
Color Colorless
Quantity 18 L
Packaging Returnable Stainless Steel Can
Molecular Formula C4H10O
Boiling Point 34°C to 35°C (1013 hPa)
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
SMILES CCOCC
IUPAC Name ethoxyethane
Molecular Weight (g/mol) 74.12
Chemical Name or Material Diethyl ether
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: diethyl ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.