missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Ethoxymethylidenemalonate, TRC

Product Code. 30379782
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30379782 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30379782 Supplier Toronto Research Chemicals Supplier No. TRCD44430025G
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl 2-ethoxymethylidene malonate
CAS 87-13-8
Molecular Formula C10 H16 O5
Quantity 25 g
Synonym Propanedioic acid, 2-(ethoxymethylene)-, 1,3-diethyl ester, Malonic acid, (ethoxymethylene)-, diethyl ester (6CI,7CI,8CI), Propanedioic acid, (ethoxymethylene)-, diethyl ester (9CI), (Ethoxymethylene)malonic acid diethyl ester, (Ethoxymethylene)propanedioic acid diethyl ester, 2-(Ethoxymethylene)malonic acid diethyl ester, 2-Ethoxy-1,1-bis(ethoxycarbonyl)ethene, Diethyl (ethoxymethylene)malonate, Diethyl (ethoxymethylene)propanedioate, Diethyl 2-(ethoxymethylene)malonate, Diethyl 2-(ethoxymethylene)propanedioate, Diethyl 2-(ethoxymethylidene)propanedioate, Diethyl 2-[(ethyloxy)methylidene]propanedioate, Diethyl 2-ethoxymethylidene malonate, Diethyl ethoxymethylidenemalonate, Diethyl[(ethyloxy)methylidene]propanedioate, Ethyl 3-ethoxy-2-(ethoxymethylene)-3-oxopropanoate, NSC 62117, NSC 9068, Nalidixic acid impurity C (EP)
InChI Formula InChI=1S/C10H16O5/c1-4-13-7-8(9(11)14-5-2)10(12)15-6-3/h7H,4-6H2,1-3H3
SMILES CCOC=C(C(=O)OCC)C(=O)OCC
IUPAC Name diethyl 2-(ethoxymethylidene)propanedioate
Molecular Weight (g/mol) 216.23
Formula Weight 216.0998
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.