missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Fumarate, TRC

Product Code. 30380996
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380996 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380996 Supplier Toronto Research Chemicals Supplier No. TRCD44449525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl fumarate
CAS 623-91-6
Molecular Formula C8 H12 O4
Quantity 25 g
Synonym 2-Butenedioic acid (2E)-, diethyl ester (9CI), 2-Butenedioic acid (E)-, diethyl ester, Fumaric acid, diethyl ester (6CI,8CI), (2E)-2-Butenedioic acid diethyl ester, (E)-But-2-enedioic acid diethyl ester, Diethyl (E)-but-2-enedioate, Diethyl fumarate, Diethyl fumaric acid ester, Ethyl fumarate, NSC 20954
InChI Formula InChI=1S/C8H12O4/c1-3-11-7(9)5-6-8(10)12-4-2/h5-6H,3-4H2,1-2H3/b6-5+
SMILES CCOC(=O)\C=C\C(=O)OCC
IUPAC Name diethyl (E)-but-2-enedioate
Molecular Weight (g/mol) 172.18
Formula Weight 172.0736
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.