missing translation for 'onlineSavingsMsg'
Learn More

Diethyl glutarate, 98%, Thermo Scientific™

Product Code. 11377656
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11377656

Brand: Thermo Scientific Alfa Aesar L11259.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 818-38-2
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.22
MDL Number MFCD00009212
InChI Key OUWSNHWQZPEFEX-UHFFFAOYSA-N
Synonym diethyl glutarate, ethyl glutarate, pentanedioic acid, diethyl ester, glutaric acid diethyl ester, glutaric acid, diethyl ester, pentanedioic acid, 1,5-diethyl ester, diethylglutarate, glutaric acid diethyl, c8-c26 alkylalcohol glutarate diester, pentanedioic acid, c18-28-alkyl esters
PubChem CID 13163
ChEBI CHEBI:87319
IUPAC Name diethyl pentanedioate
SMILES CCOC(=O)CCCC(=O)OCC

Specifications

Density 1.022
Boiling Point 236°C to 237°C
Flash Point 96°C (204°F)
Refractive Index 1.423
Quantity 25 g
Beilstein 1777327
Formula Weight 188.23
Percent Purity 98%
Chemical Name or Material Diethyl glutarate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.