missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Hexyl Phosphate-d11, TRC
Specifications
Specifications
| Chemical Name or Material | Diethyl Hexyl Phosphate-d11 |
| Molecular Formula | C10 D11 H12 O4 P |
| Quantity | 25 mg |
| Synonym | Phosphoric Acid Diethyl Hexyl Ester-d11, Ethyl Hexyl Phosphate-d11 |
| InChI Formula | InChI=1S/C10H23O4P/c1-4-7-8-9-10-14-15(11,12-5-2)13-6-3/h4-10H2,1-3H3/i1D3,4D2,7D2,8D2,9D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])COP(=O)(OCC)OCC |
| IUPAC Name | diethyl 2,2,3,3,4,4,5,5,6,6,6-undecadeuteriohexyl phosphate |
| Molecular Weight (g/mol) | 249.329 |
| Formula Weight | 249.202 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?